Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RTF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552330
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552330 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552330 Supplier Invitrogen™ Supplier No. PA552330
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GNHNSKPVYR VAEITGVVET AKVYQLGGTR TNKGLQLRHG NDQRVFRLEF VSNQEFTESE FMKWKEAMFS AGMQLPTLDE INKKELSIKE ALNYKFNDQD IEEIVKEKER FRKAPPNYAM KKTQL Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 99%.

Three isoforms of serine/threonine kinases, designated alphaPAK p68, betaPAK p65 and gammaPAK p62, have been shown to exhibit a high degree of sequence homology with the S. cerevisiae kinase Ste 20, involved in pheromone signaling. The alpha, beta and gammaPAK isoforms complex specifically with Rac1 and Cdc42 in their active GTP-bound state, inhibiting their intrinsic GTPase activity leading to their autophosphorylation. There are eight sites of autophosphorylation on gammaPAK, including Ser 19, Ser 141 and Thr 402, and phosphorylation of Ser 141 and Thr 402 is correlated with gammaPAK activation. Once phosphorylated and their affinity for Rac/Cdc42 reduced, the PAK isoforms disassociate from the complex to seek downstream substrates.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RTF1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RTF1
Gene Accession No. Q92541
Gene Alias 2900005O08Rik; 6530416A09Rik; AI853581; AU043144; AW553985; cbp82; CG10955; CG10955-PA; CG10955-PB; Dmel\CG10955; Dmel_CG10955; dRtf1; gene trap locus 7; Gtl7; KIAA0252; ortholog of mouse gene trap locus 7; RNA polymerase-associated protein Rtf1; RNA polymerase-associated protein RTF1 homolog; Rtf1; RTF1 homolog, Paf1/RNA polymerase II complex component; Rtf1 Paf1/RNA polymerase II complex component homolog; Rtf1, Paf1/RNA polymerase II complex component, homolog; Rtf1, Paf1/RNA polymerase II complex component, homolog (S. cerevisiae); Rtf1-PA; Rtf1-PB
Gene Symbols RTF1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RTF1. Recombinant protein control fragment (Product #RP-89460).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23168
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.