Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RTF1 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS12261R

Description
Three isoforms of serine/threonine kinases, designated alphaPAK p68, betaPAK p65 and gammaPAK p62, have been shown to exhibit a high degree of sequence homology with the S. cerevisiae kinase Ste 20, involved in pheromone signaling. The alpha, beta and gammaPAK isoforms complex specifically with Rac1 and Cdc42 in their active GTP-bound state, inhibiting their intrinsic GTPase activity leading to their autophosphorylation. There are eight sites of autophosphorylation on gammaPAK, including Ser 19, Ser 141 and Thr 402, and phosphorylation of Ser 141 and Thr 402 is correlated with gammaPAK activation. Once phosphorylated and their affinity for Rac/Cdc42 reduced, the PAK isoforms disassociate from the complex to seek downstream substrates.Specifications
RTF1 | |
Polyclonal | |
Unconjugated | |
RTF1 | |
2900005O08Rik; 6530416A09Rik; AI853581; AU043144; AW553985; cbp82; CG10955; CG10955 gene product from transcript CG10955-RB; CG10955-PA; CG10955-PB; Dmel\CG10955; Dmel_CG10955; dRtf1; gene trap locus 7; GTL7; KIAA0252; ortholog of mouse gene trap locus 7; RNA polymerase-associated protein Rtf1; RNA polymerase-associated protein RTF1 homolog; RTF1; RTF1 homolog, Paf1/RNA polymerase II complex component; Rtf1 Paf1/RNA polymerase II complex component homolog; Rtf1, Paf1/RNA polymerase II complex component, homolog; Rtf1, Paf1/RNA polymerase II complex component, homolog (S. cerevisiae); Rtf1-PA; Rtf1-PB | |
Rabbit | |
Protein A | |
RUO | |
23168 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q92541 | |
RTF1 | |
KLH conjugated synthetic peptide derived from human RTF1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction