Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RTL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140010
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5140010 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5140010 Supplier Invitrogen™ Supplier No. PA5140010
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: DRLTVLLPGH WVFFFSHFNF DVMELPEQDG GRALPPVRNL RWRRAFQRNT AARQTLLLAS RGFPRDPSTE SGEEENEEQD ELNEQILR Highest antigen sequence indentity to the following orthologs: Mouse - 48%.

Plays an essential role in capillaries endothelial cells for the maintenance of feto-maternal interface and for development of the placenta.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RTL1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RTL1
Gene Accession No. A6NKG5
Gene Alias 1-Mar; Mammalian retrotransposon derived protein 1; Mar1; MART1; Paternally expressed gene 11 protein; PEG11; Retrotransposon-derived protein PEG11; retrotransposon-like 1; retrotransposon-like protein 1; RTL1
Gene Symbols RTL1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RTL1. Recombinant protein control fragment (Product #RP-109635).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 388015
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.