Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RTL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA523513
Description
RTL1 Polyclonal Antibody for Western Blot
Plays an essential role in capillaries endothelial cells for the maintenance of feto-maternal interface and for development of the placenta.
Specifications
| RTL1 | |
| Polyclonal | |
| Unconjugated | |
| RTL1 | |
| 1-Mar; Mammalian retrotransposon derived protein 1; Mar1; MART1; Paternally expressed gene 11 protein; PEG11; Retrotransposon-derived protein PEG11; retrotransposon-like 1; retrotransposon-like protein 1; RTL1 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 388015 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| A6NKG5 | |
| RTL1 | |
| KLH conjugated synthetic peptide between 1330-1358 amino acids from the C-terminal region of human RTL1. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction