Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RTN1-A/NSP Antibody (MON161), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP197678MFV500
Description
RTN1-A/NSP Monoclonal antibody specifically detects RTN1-A/NSP in Human, Mouse, Rat, Hamster samples. It is validated for Flow CytometrySpecifications
RTN1-A/NSP | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Hamster | |
Purified |
Flow Cytometry | |
MON161 | |
Flow Cytometry | |
NSP | |
Derived by fusion of Mouse myeloma cells with spleen cells from a Mouse immunized with a partially purified bacterially expressed Reticulon-1A (NSP-A) hybrid protein. | |
0.1 mL | |
Neuroscience | |
6252 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction