Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RTN1-A/NSP Antibody (MON161), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP197678PE
Description
RTN1-A/NSP Monoclonal specifically detects RTN1-A/NSP in Human, Mouse, Rat, Hamster samples. It is validated for Flow Cytometry.Specifications
RTN1 | |
Monoclonal | |
PE | |
Neuroendocrine-specific proteinNSPMGC133250, reticulon 1, reticulon-1 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
MON-161 exclusively recognizes the 135 kD Reticulon-1A protein in immunoblots of NCI-H82 and other SCLC cell lines, and stains normal and pathological neural and neuroendocrine tissues. | |
Store at 4C in the dark. |
Flow Cytometry | |
MON161 | |
Flow Cytometry | |
RTN1 | |
MON-161 is a Mouse monoclonal IgG1 antibody derived by fusion of Mouse myeloma cells with spleen cells from a Mouse immunized with a partially purified bacterially expressed Reticulon-1A (NSP-A) hybrid protein (betaGAL-NSP-A 6-776). The epitope of MON-161 | |
0.1 mL | |
Neuroscience | |
6252 | |
Human, Mouse, Rat, Hamster | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction