Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RTN1-A/NSP Antibody (MON162), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP197677AF750
Description
RTN1-A/NSP Monoclonal specifically detects RTN1-A/NSP in Human, Mouse, Rat, Hamster samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
RTN1-A/NSP | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
Mouse | |
Protein A or G purified | |
Neuroscience | |
6252.0 | |
Human, Mouse, Rat, Hamster | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
MON162 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
RTN1 | |
MON-162 is a Mouse monoclonal IgG1 antibody derived by fusion of Mouse myeloma cells with spleen cells from a Mouse immunized with a partially purified bacterially expressed Reticulon-1A (NSP-A) hybrid protein (beta-GAL-NSP-A 6-776). The epitope of MON-16 | |
0.1 mL | |
Primary | |
MON-162 exclusively recognizes the 135 kD Reticulon-1A protein in immunoblots of NCI-H82 and other SCLC cell lines, and stains normal and pathological neural and neuroendocrine tissues. | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction