Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RTN1-A/NSP Antibody (RNL-4), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP197673AF488
Description
RTN1-A/NSP Monoclonal specifically detects RTN1-A/NSP in Human, Rat, Porcine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
RTN1 | |
Monoclonal | |
Alexa Fluor 488 | |
Neuroendocrine-specific proteinNSPMGC133250, reticulon 1, reticulon-1 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
RNL-4 reacts with peripheral nerves and ganglia of various tissues and cross-reacts with smooth muscle cells and myoepithelium. In the central nervous system it reacts with the neurohypophysis and pars intermedia of the pituitary gland, and a weak diffuse | |
Store at 4C in the dark. |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
RNL-4 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen | |
RTN1 | |
RNL-4 is a Mouse monoclonal IgG1 antibody derived by fusion of SP2/0-Ag14 Mouse myeloma cells, with spleen cells from a BALB/c Mouse immunized with a synthetic peptide encompassing the first 20 amino acids on the N-terminal of Reticulon-1C (NSP-C). | |
0.1 mL | |
Neuroscience | |
6252 | |
Human, Rat, Rat (Negative) | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction