Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RTN1 Monoclonal Antibody (MON160)
GREENER_CHOICE

Catalog No. MA106810
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA106810 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA106810 Supplier Invitrogen™ Supplier No. MA106810
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-06810 detects reticulon-1A (NSP-A) in human, mouse, hamster, and rat samples. MA1-06810 has sucessfully been used in Western blotting, FACS, ICC and immunohistochemistry procedures. By Western blot it detects a ~135 kDa band representing reticulon-1A. The MA1-06810 immunogen is a partially purified bacterially expressed reticulon-1A (NSP-A) hybrid protein.

Reticulon-1 was formerly designated NSP for Neuroendocrine-Specific-Protein, because it is specifically expressed in neural and neuroendocrine tissues. The NSP-gene has been mapped by fluorescence in situ hybridization to human chromosome 14q21-q22. The NSP-gene encodes three overlapping proteins, i.e. reticulon-1A (NSP-A), reticulon-1B (NSP-B), and reticulon-1C (NSP-C). These proteins were found to be anchored to membranes of the endoplasmic reticulum through their common carboxy-terminal regions. Reticulon-1A is a protein with a molecular weight of approximately 135 kDa, which occurs in various isoforms presumably depending on the degree of phosphorylation of serine residues. In lung cancer diagnosis Reticulon-1A appeared to be a reliable marker for the detection of neuroendocrine differentiation, since most of the small cell lung carcinoma (SCLC) and carcinoid tumors showed expression of Reticulon-1A. Reticulon-1B is a phosphoprotein with a MW of 45 kDa and is restricted to the lung cancer cell line NCI-H82. Reticulon-1B is so far not found in human tissues. Reticulon-1C is a protein with a MW of 23 kDa which is not phosphorylated and is found with Reticulon-1A in SCLC (cell lines) and not in non-SCLC (cell cultures).
TRUSTED_SUSTAINABILITY

Specifications

Antigen RTN1
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone MON160
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene RTN1
Gene Accession No. Q16799, Q64548, Q8K0T0
Gene Alias 0710005K15Rik; 4930441F12Rik; I79_024631; LOC100750648; MGC133250; MGC40985; Neuroendocrine-specific protein; Nsp; R75279; reticulon 1; reticulon-1; RP23-450G14.1; RTN1; Rtn1-a; Rtn1-b; Rtn1-c; S-rex
Gene Symbols LOC100750648, RTN1
Host Species Mouse
Immunogen Partially purified bacterially expressed reticulon-1A (NSP-A) hybrid protein.
Purification Method Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100750648, 104001, 116644, 6252
Target Species Human, Mouse, Rat, Hamster
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.