Learn More
Invitrogen™ RTN1 Monoclonal Antibody (RNL-4)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA106815
Description
MA1-06815 detects reticulon-1C(NSP-C) in human, porcine, and rat samples. MA1-06815 has sucessfully been used in flow cytometry, Western blotting, immunocytochemistry and immunohistochemistry procedures. By Western blot it detects a ~23 kDa band representing reticulon-1C. The MA1-06815 immunogen is a synthetic peptide encompassing the unique 20 N-terminal amino acid sequence of reticulon-1C.
Reticulon-1 was formerly designated NSP for Neuroendocrine-Specific-Protein, because it is specifically expressed in neural and neuroendocrine tissues. The NSP-gene has been mapped by fluorescence in situ hybridization to human chromosome 14q21-q22. The NSP-gene encodes three overlapping proteins, i.e. reticulon-1A (NSP-A), reticulon-1B (NSP-B), and reticulon-1C (NSP-C). These proteins were found to be anchored to membranes of the endoplasmic reticulum through their common carboxy-terminal regions. Reticulon-1A is a protein with a molecular weight of approximately 135 kDa, which occurs in various isoforms presumably depending on the degree of phosphorylation of serine residues. In lung cancer diagnosis Reticulon-1A appeared to be a reliable marker for the detection of neuroendocrine differentiation, since most of the small cell lung carcinoma (SCLC) and carcinoid tumors showed expression of Reticulon-1A. Reticulon-1B is a phosphoprotein with a MW of 45 kDa and is restricted to the lung cancer cell line NCI-H82. Reticulon-1B is so far not found in human tissues. Reticulon-1C is a protein with a MW of 23 kDa which is not phosphorylated and is found with Reticulon-1A in SCLC (cell lines) and not in non-SCLC (cell cultures).
Specifications
RTN1 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
Q16799, Q64548 | |
RTN1 | |
Synthetic peptide encompassing the unique 20 N-terminal amino acid sequence of reticulon-1C. | |
100 μg | |
Primary | |
Human, Rat, Pig | |
Antibody | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry | |
RNL-4 | |
Unconjugated | |
RTN1 | |
0710005K15Rik; 4930441F12Rik; I79_024631; LOC100750648; MGC133250; MGC40985; Neuroendocrine-specific protein; Nsp; R75279; reticulon 1; reticulon-1; RP23-450G14.1; RTN1; Rtn1-a; Rtn1-b; Rtn1-c; S-rex | |
Mouse | |
Purified | |
RUO | |
100152394, 116644, 6252 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.