Learn More
Invitrogen™ RTN1 Polyclonal Antibody

Description
Immunogen sequence: TGVAGVSSAM DHTFSTTSKD GEGSCYTSLI SDICYPPQED STYFTGILQK ENGHVTISES PEELGTPGPS LPDVPGIESR GLFSSDSGIE MTPAESTEVN KILADPLDQM KAEAYKYIDI TRPEEVKHQE QHHPELEDKD LDFKNKDTDI SIKPEGVREP DKPAPVEGKI IKDHLLEEST FAPYIDDLSE EQRRAPQITT PVKITLTEIE PSVETTTQEK TPEKQDICLK PSPDTVPTVT VSEPEDDSPG SITPPSSGTE P; Positive Samples: Mouse lung; Cellular Location: Endoplasmic reticulum membrane, Multi-pass membrane protein.
Specifications
Specifications
| Antigen | RTN1 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.53 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | RTN1 |
| Gene Accession No. | Q8K0T0 |
| Gene Alias | 0710005K15Rik; 4930441F12Rik; I79_024631; LOC100750648; MGC133250; MGC40985; Neuroendocrine-specific protein; Nsp; R75279; reticulon 1; reticulon-1; RP23-450G14.1; RTN1; Rtn1-a; Rtn1-b; Rtn1-c; S-rex |
| Gene Symbols | RTN1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.