Learn More
Invitrogen™ RTN2 Monoclonal Antibody (6A11)

Description
Peptide Sequence: MGQVLPVFAH CKEAPSTASS TPDSTEGGND DSDFRELHTA REFSEEDEEE TTSQDWGTPR ELTFSYIAFD GVVGSGGRRD STARRPRPQG RSVSEPRDQH.
Specifications
Specifications
| Antigen | RTN2 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 6A11 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | RTN2 |
| Gene Accession No. | O75298 |
| Gene Alias | formerly RTN2w; MMS10-P; Ms10p; neuroendocrine-specific protein-like 1; Neuroendocrine-specific protein-like I; NSP2; Nspl1; NSPLI; NSP-like 1; NSP-like protein 1; NSP-like protein I; reticulon 2; reticulon 2 (Z-band associated protein); reticulon-2; RTN2; RTN2-B; SPG12 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.