Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RTN2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5145194
Description
Immunogen sequence: LRLRLDHLGW VARGTGSGED SSTSSSTPLE DEEPQEPNRL ETGEAGEELD LRLRLAQPSS PEVLTPQLSP GSG Antigen sequence identity: Mouse 69%; Rat 67%
RTN2 Polyclonal antibody specifically detects RTN2 in Human samples. It is validated for Immunocytochemistry, Western Blot
Specifications
| RTN2 | |
| Polyclonal | |
| Unconjugated | |
| RTN2 | |
| formerly RTN2w, MMS10-P, Ms10p, neuroendocrine-specific protein-like 1, Neuroendocrine-specific protein-like I, NSP2, Nspl1, NSPLI, NSP-like 1, NSP-like protein 1, NSP-like protein I, reticulon 2, reticulon 2 (Z-band associated protein), reticulon-2, RTN2, RTN2-B, SPG12 | |
| Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 140-212. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunocytochemistry, Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O75298 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 6253 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction