Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RTN3 Monoclonal Antibody (1E11)
GREENER_CHOICE

Catalog No. PIMA515538
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515538 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515538 Supplier Invitrogen™ Supplier No. MA515538
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15538 targets RTN3 in indirect ELISA, IF applications and shows reactivity with Human samples. The MA5-15538 immunogen is purified recombinant fragment of RTN3 expressed in E. Coli. MA5-15538 detects RTN3 which has a predicted molecular weight of approximately 113kDa.

RTN3 is a member of reticulons, which are associated with the endoplasmic reticulum, and are involved in neuroendocrine secretion or in membrane trafficking in neuroendocrine cells. Alternatively spliced transcript variants encoding different isoforms have been identified. Multiple promoters rather than alternative splicing of internal exons seem to be involved in this diversity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RTN3
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone 1E11
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.03% sodium azide
Gene RTN3
Gene Accession No. O95197
Gene Alias ASY interacting protein; ASYIP; HAP; Homolog of ASY protein; neuroendocrine-specific protein-like 2; Neuroendocrine-specific protein-like II; nsp-c; NSPL2; NSPLII; Nsp-like II; NSP-like protein 2; NSP-like protein II; reticulon 3; reticulon-3; RTN3; RTN3-A1
Gene Symbols RTN3
Host Species Mouse
Immunogen Purified recombinant fragment of RTN3 expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10313
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.