Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RTN3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553360
Description
Immunogen sequence: IMTSSFLSSS EIHNTGLTIL HGEKSHVLGS QPILAKEGKD HLDLLDMKKM EKPQGTSNNV SDSSVSLAAG VHCDRPSIPA SFPEHPAFLS KKIGQVEEQI DKETKNPNGV SSREAKTALD ADDRFTLLTA QKPPTEY Highest antigen sequence identity to the following orthologs: Mouse - 53%, Rat - 47%.
RTN3 is a member of reticulons, which are associated with the endoplasmic reticulum, and are involved in neuroendocrine secretion or in membrane trafficking in neuroendocrine cells. Alternatively spliced transcript variants encoding different isoforms have been identified. Multiple promoters rather than alternative splicing of internal exons seem to be involved in this diversity.
Specifications
| RTN3 | |
| Polyclonal | |
| Unconjugated | |
| RTN3 | |
| ASY interacting protein; ASYIP; HAP; Homolog of ASY protein; neuroendocrine-specific protein-like 2; Neuroendocrine-specific protein-like II; nsp-c; NSPL2; NSPLII; Nsp-like II; NSP-like protein 2; NSP-like protein II; reticulon 3; reticulon-3; RTN3; RTN3-A1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10313 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O95197 | |
| RTN3 | |
| Recombinant protein corresponding to Human RTN3. Recombinant protein control fragment (Product #RP-90846). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction