Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RTN3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578316
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578316 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578316 Supplier Invitrogen™ Supplier No. PA578316
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: HeLa, HeLa membrane fraction extract Predicted Reactivity: Chimpanzee (98%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

RTN3 is a member of reticulons, which are associated with the endoplasmic reticulum, and are involved in neuroendocrine secretion or in membrane trafficking in neuroendocrine cells. Alternatively spliced transcript variants encoding different isoforms have been identified. Multiple promoters rather than alternative splicing of internal exons seem to be involved in this diversity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RTN3
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.69 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene RTN3
Gene Accession No. O95197
Gene Alias ASY interacting protein; ASYIP; HAP; Homolog of ASY protein; neuroendocrine-specific protein-like 2; Neuroendocrine-specific protein-like II; nsp-c; NSPL2; NSPLII; Nsp-like II; NSP-like protein 2; NSP-like protein II; reticulon 3; reticulon-3; RTN3; RTN3-A1
Gene Symbols RTN3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human RTN3. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10313
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.