Learn More
RTN4IP1 Mouse anti-Human, Clone: OTI2A9, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF502623

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a novel mitochondrial protein that interacts with reticulon 4, which is a potent inhibitor of regeneration following spinal cord injury. The interaction of reticulon 4 with mitochondrial proteins may provide insight into the mechanisms for reticulon-induced inhibition of neurite growth.Specifications
RTN4IP1 | |
Monoclonal | |
Unconjugated | |
RTN4IP1 | |
NIMP, OPA10 | |
Mouse | |
Affinity Chromatography | |
RUO | |
84816 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Western Blot | |
OTI2A9 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q8WWV3 | |
RTN4IP1 | |
Full length human recombit protein of human RTN4IP1 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.