Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RTN4IP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531794
Description
Recommended positive controls: mouse heart. Predicted reactivity: Mouse (96%), Rat (95%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a novel mitochondrial protein that interacts with reticulon 4, which is a potent inhibitor of regeneration following spinal cord injury. The interaction of reticulon 4 with mitochondrial proteins may provide insight into the mechanisms for reticulon-induced inhibition of neurite growth.
Specifications
RTN4IP1 | |
Polyclonal | |
Unconjugated | |
Rtn4ip1 | |
D10Ertd690e; Nimp; NOGO-interacting mitochondrial protein; OPA10; reticulon 4 interacting protein 1; reticulon 4 interacting protein 1, mitochondrial; reticulon-4-interacting protein 1, mitochondrial; RGD1563384; RTN4IP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
170728, 84816 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.64 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8WWV3, Q924D0 | |
Rtn4ip1 | |
Recombinant fragment corresponding to a region within amino acids 6 and 222 of Human RTN4IP1. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction