Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RTN4IP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557625
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557625 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557625 Supplier Invitrogen™ Supplier No. PA557625
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TLVTPFLLNM DRLGIADGML QTGVTVGSKA LKHFWKGVHY RWAFFMASGP CLDDIAELVD AGKIRPVIEQ TFPFSKVPEA FLKVERGHAR GK Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.

This gene encodes a novel mitochondrial protein that interacts with reticulon 4, which is a potent inhibitor of regeneration following spinal cord injury. The interaction of reticulon 4 with mitochondrial proteins may provide insight into the mechanisms for reticulon-induced inhibition of neurite growth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RTN4IP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Rtn4ip1
Gene Accession No. Q8WWV3
Gene Alias D10Ertd690e; Nimp; NOGO-interacting mitochondrial protein; OPA10; reticulon 4 interacting protein 1; reticulon 4 interacting protein 1, mitochondrial; reticulon-4-interacting protein 1, mitochondrial; RGD1563384; RTN4IP1
Gene Symbols Rtn4ip1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RTN4IP1. Recombinant protein control fragment (Product #RP-94819).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84816
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.