Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RTN4IP1 Monoclonal Antibody (OTI1E1)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA525452
Description
RTN4IP1 Monoclonal Antibody for Western Blot, Flow
This gene encodes a novel mitochondrial protein that interacts with reticulon 4, which is a potent inhibitor of regeneration following spinal cord injury. The interaction of reticulon 4 with mitochondrial proteins may provide insight into the mechanisms for reticulon-induced inhibition of neurite growth.
Specifications
RTN4IP1 | |
Monoclonal | |
0.65 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8WWV3 | |
Rtn4ip1 | |
Full length human recombinant protein of RTN4IP1 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Western Blot | |
OTI1E1 | |
Unconjugated | |
Rtn4ip1 | |
D10Ertd690e; Nimp; NOGO-interacting mitochondrial protein; OPA10; reticulon 4 interacting protein 1; reticulon 4 interacting protein 1, mitochondrial; reticulon-4-interacting protein 1, mitochondrial; RGD1563384; RTN4IP1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
84816 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction