Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RTN4RL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110335
Description
Immunogen sequence: LRRLTTLFLF NNSLSELQGE CLAPLGALEF LRLNGNPWDC GCRARSLWEW LQRFRGSSSA VPCVSPGLRH GQDLKLLRAE DFRNCTGPAS PHQIKSHTLT TTDRAARKEH HSPHGPTRSK GHPHGPRPGH RKPGKNCTNP RNRNQISKAG AGKQAPELPD YAPDYQHKFS FDIMPTARPK RKGKCARRTP IRAPSGVQQA S.
RTN4RL1 may play a role in regulating axonal regeneration and plasticity in the adult central nervous system.
Specifications
| RTN4RL1 | |
| Polyclonal | |
| Unconjugated | |
| Rtn4rl1 | |
| NgR3; Ngrh2; NGRL2; Nogo receptor-like 2; Nogo-66 receptor homolog 2; Nogo-66 receptor-related protein 3; reticulon 4 receptor like 1; reticulon 4 receptor-like 1; reticulon-4 receptor-like 1; RGD:727812}; Rtn4rl1; rtn4rl1 {ECO:0000312 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 237847, 303311 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.03 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q80WD0, Q8K0S5 | |
| Rtn4rl1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 220-420 of human RTN4RL1 (NP_848663.1). | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction