Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RTN4RL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA525189
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA525189 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA525189 Supplier Invitrogen™ Supplier No. PA525189
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RTN4RL1 Polyclonal Antibody for Western Blot, IHC (P), Flow

RTN4RL1 may play a role in regulating axonal regeneration and plasticity in the adult central nervous system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RTN4RL1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.50 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Rtn4rl1
Gene Accession No. Q86UN2
Gene Alias NgR3; Ngrh2; NGRL2; Nogo receptor-like 2; Nogo-66 receptor homolog 2; Nogo-66 receptor-related protein 3; reticulon 4 receptor like 1; reticulon 4 receptor-like 1; reticulon-4 receptor-like 1; RGD:727812}; Rtn4rl1; rtn4rl1 {ECO:0000312
Gene Symbols Rtn4rl1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 326-355 amino acids from the C-terminal region of human RTN4RL1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146760
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.