Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rtn4rl2, anti-Rat, Clone: P14, Abnova™
Monoclonal Antibody
Supplier: Abnova Corporation RAB00092
Specifications
Rtn4rl2 | |
Monoclonal | |
Unconjugated | |
In PBS, pH 7.2 (0.02% Proclin 300). | |
Ngrh1 | |
Original antibody is raised against His6 tagged fragment corresponding to amino acids 279-420 of rat Rtn4rl2. | |
RUO | |
311169 | |
Human, Rat | |
Antibody | |
IgG κ |
ELISA, Flow Cytometry, Immunoprecipitation, Western Blot | |
P14 | |
ELISA (0.1 ug/mL) Flow Cytometry (1 ug/mL) Immunoprecipitation (10 ug/mL) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
reticulon 4 receptor-like 2 | |
Rtn4rl2 | |
200 μg | |
Primary | |
This antibody recognizes human NgR2 with a Kd of 2.3 nM and to rat NgR2 with a Kd of 0.48 nM (bivalent binding). | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction