Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RTN4RL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598577
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598577 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598577 Supplier Invitrogen™ Supplier No. PA598577
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RTN4RL2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

Reticulon 4 receptor-like 2 (RTN4RL2), also known as NgR2and NgRH1, is a 60 kDa glycosylphosphatidylinositol-anchored protein that is expressed primarily in the brain. It contains eight leucine-rich repeats that are flanked by cysteine-rich sequences at both the N- and C-termini. The membrane anchored RTN4RL2 can be solubilized through the action of phospholipase or an unidentified MTMMP to generate a 46 kDa soluble receptor. It has been suggested that RTN4RL2 may play a role in regulating axonal regeneration and plasticity in the adult central nervous system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RTN4RL2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.12 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene RTN4RL2
Gene Accession No. Q86UN3
Gene Alias ngR2; NGRH1; NGRL3; Nogo receptor-like 3; Nogo-66 receptor homolog 1; Nogo-66 receptor-related protein 2; reticulon 4 receptor-like 2; reticulon-4 receptor-like 2; RGD:727797}; Rtn4rl2; rtn4rl2 {ECO:0000312
Gene Symbols RTN4RL2
Host Species Rabbit
Immunogen Peptide sequence from Human Reticulon-4 receptor-like 2 protein (355-372AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 349667
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.