Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RTN4RL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534488
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534488 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534488 Supplier Invitrogen™ Supplier No. PA534488
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is rat brain tissue lysate. PA5-34488 can be used with blocking peptide PEP-1530.

Reticulon 4 receptor-like 2 (RTN4RL2), also known as NgR2and NgRH1, is a 60 kDa glycosylphosphatidylinositol-anchored protein that is expressed primarily in the brain. It contains eight leucine-rich repeats that are flanked by cysteine-rich sequences at both the N- and C-termini. The membrane anchored RTN4RL2 can be solubilized through the action of phospholipase or an unidentified MTMMP to generate a 46 kDa soluble receptor. It has been suggested that RTN4RL2 may play a role in regulating axonal regeneration and plasticity in the adult central nervous system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RTN4RL2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene RTN4RL2
Gene Accession No. Q80WD1, Q86UN3
Gene Alias ngR2; NGRH1; NGRL3; Nogo receptor-like 3; Nogo-66 receptor homolog 1; Nogo-66 receptor-related protein 2; reticulon 4 receptor-like 2; reticulon-4 receptor-like 2; RGD:727797}; Rtn4rl2; rtn4rl2 {ECO:0000312
Gene Symbols RTN4RL2
Host Species Rabbit
Immunogen An 18 amino acid peptide near the carboxy terminus of human RTN4RL2.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 311169, 349667
Target Species Human, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.