Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RTP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA524028
Description
This antibody is predicted to react with non-human primate based on sequence homology.
Specifically promotes functional cell surface expression of olfactory receptors, but not of other GPCRs.
Specifications
RTP1 | |
Polyclonal | |
Unconjugated | |
Rtp1 | |
3CxxC-type zinc finger protein 1; Gm1766; Gm604; receptor (chemosensory) transporter protein 1; receptor transporter protein 1; receptor transporting protein 1; receptor-transporting protein 1; RGD1560980; Rtp1; Z3CXXC1; zinc finger, 3CxxC-type 1 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
132112, 239766 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.25 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P59025, Q8C8C1 | |
Rtp1 | |
KLH conjugated synthetic peptide between 161-189 amino acids from the C-terminal region of human RTP1. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction