Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RTP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555448
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555448 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555448 Supplier Invitrogen™ Supplier No. PA555448
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YRIHVASRPD SGPHRAEFCE ACQEGIVHWK PSEKLLEEEV TTYTSEASKP RAQAGSGYNF Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 72%.

RTP2 specifically promotes functional cell surface expression of olfactory receptors, but not of other GPCRs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RTP2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RTP2
Gene Accession No. Q5QGT7
Gene Alias 3CxxC-type zinc finger protein 2; Gm605; receptor (chemosensory) transporter protein 2; receptor transporter protein 2; receptor transporting protein 2; receptor-transporting protein 2; Rtp2; Z3CXXC2; zinc finger, 3CxxC-type 2
Gene Symbols RTP2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RTP2. Recombinant protein control fragment (Product #RP-90583).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 344892
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.