Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RTP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112997
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112997 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112997 Supplier Invitrogen™ Supplier No. PA5112997
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RTP2 Polyclonal Antibody for Western Blot, ELISA

RTP2 specifically promotes functional cell surface expression of olfactory receptors, but not of other GPCRs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RTP2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene RTP2
Gene Accession No. Q5QGT7, Q80ZI2
Gene Alias 3CxxC-type zinc finger protein 2; Gm605; receptor (chemosensory) transporter protein 2; receptor transporter protein 2; receptor transporting protein 2; receptor-transporting protein 2; Rtp2; Z3CXXC2; zinc finger, 3CxxC-type 2
Gene Symbols RTP2
Host Species Rabbit
Immunogen Recombinant Human Receptor-transporting protein 2 protein (1-196AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 224055, 344892
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.