Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RTP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564190
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564190 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564190 Supplier Invitrogen™ Supplier No. PA564190
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CSWSQYEMPE FSSDSTMRIL SNLVQHILKK YYGNGTRKSP EMPVILEVSL EGSHDTANCE ACTLGICG Highest antigen sequence identity to the following orthologs: Mouse - 46%, Rat - 48%.

RTP4 belongs to the TMEM7 family. The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RTP4
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RTP4
Gene Accession No. Q96DX8
Gene Alias 28 kDa interferon-responsive protein; 28kD interferon alpha responsive protein; 28kD interferon responsive protein; 3CxxC-type zinc finger protein 4; 5830458K16Rik; IFRG28; receptor (chemosensory) transporter protein 4; receptor transporter protein 4; receptor transporting protein 4; receptor-transporting protein 4; Rtp4; Z3CXXC4; zinc finger, 3CxxC-type 4
Gene Symbols RTP4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RTP4. Recombinant protein control fragment (Product #RP-103578).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64108
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.