Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RTP4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA524039
Description
RTP4 Polyclonal Antibody for Western Blot, Flow
RTP4 belongs to the TMEM7 family. The function of this protein remains unknown.
Specifications
RTP4 | |
Polyclonal | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q96DX8 | |
RTP4 | |
KLH conjugated synthetic peptide between 186-215 amino acids from the C-terminal region of human RTP4. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Flow Cytometry, Western Blot | |
Unconjugated | |
RTP4 | |
28 kDa interferon-responsive protein; 28kD interferon alpha responsive protein; 28kD interferon responsive protein; 3CxxC-type zinc finger protein 4; 5830458K16Rik; IFRG28; receptor (chemosensory) transporter protein 4; receptor transporter protein 4; receptor transporting protein 4; receptor-transporting protein 4; Rtp4; Z3CXXC4; zinc finger, 3CxxC-type 4 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
64108 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction