Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RTP5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571016
Description
This target displays homology in the following species: Human: 100%.
RTP5 gene ontology annotations related to this gene include detection of chemical stimulus involved in sensory perception of bitter taste; protein insertion into membrane; protein targeting to membrane.
Specifications
RTP5 | |
Polyclonal | |
Unconjugated | |
RTP5 | |
3CxxC-type zinc finger protein 5; C2orf85; CXXC finger protein 11; CXXC11; CXXC-type zinc finger protein 11; receptor (chemosensory) transporter protein 5 (putative); receptor transporter protein 5 (putative); receptor-transporting protein 5; RTP5; Z3CXXC5; zinc finger, 3CxxC-type 5 | |
Rabbit | |
Affinity chromatography | |
RUO | |
285093 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q14D33 | |
RTP5 | |
synthetic peptide directed towards the C-terminal region of human RTP5. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction