Learn More
Description
- Removes all residual catalysts and low molecular weight fragments from the Rtx™-5 polymer
- Provides a tight, unimodal distribution
- Extremely low bleed
- For Drugs, Solvent Impurities, Pesticides, Hydrocarbons, PCB Congeners (e.g., Aroclor Mixes), Essential Oils, Semivolatiles
Specifications
Specifications
| For Use With (Application) | Drugs, Solvent Impurities, Pesticides, Hydrocarbons, PCB Congeners (e.g., Aroclor Mixes), Essential Oils, Semivolatiles |
| Film Thickness | 3 μm |
| Length (Metric) | 60 m |
| Polarity | Low Polarity |
| Coating | Fused Silica |
| Quantity | 1/Pk. |
| USP Type | G27, G36 |
| Stationary Phase | Rtx™-5 |
| Type | Capillary Column |
| Diameter (Metric) Inner | 0.53 mm |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
