Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ RU 24969 hemisuccinate
GREENER_CHOICESDP

Catalog No. 091250 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-125-0 50 mg
09-121-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 09-125-0 Supplier Tocris Bioscience™ Supplier No. 0912/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

5-HT1A and 5-HT1B agonist

RU 24969 hemisuccinate is a potent 5-HT1A/1B and moderate 5-HT2C agonist that may also release 5-HT. Centrally active following systemic administration.

Specifications

Chemical Name or Material 5-Methoxy-3-(1,2,5,6-tetrahydro-4-pyridinyl)-1H-indole hemisuccinate
CAS 66611-27-6
Assay Percent Range >99%
Quantity 50 mg
Solubility Soluble to 50mM in DMSO
Molecular Formula C14H16N2O.½C4H6O4
Formula Weight Observed MW: 287.34

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.