Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RU 28318, potassium salt, Tocris Bioscience™

Catalog No. 167250
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-725-0 50 mg
16-721-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 16-725-0 Supplier R&D Systems Supplier No. 1672/50

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Potent, selective mineralocorticoid receptor antagonist

Potent and selective antagonist for the mineralocorticoid receptor (MR). Inhibits aldosterone production and secretion, and selectively decreases ex-vivo MR binding in the hippocampus of adrenalectomised rats. Also decreases blood pressure in female rats following central administration in vivo.

Chemical Identifiers

CAS 76676-34-1
Molecular Formula C25H37KO4
Molecular Weight (g/mol) 440.665
InChI Key HXJITUGMCJCKCE-UYOQDFFISA-M
Synonym 7a,17a-17-hydroxy-3-oxo-7-propylpregn-4-ene-21-carboxylic acid, potassium salt
PubChem CID 23677972
IUPAC Name potassium;3-[(7R,8R,9S,10R,13S,14S,17R)-17-hydroxy-10,13-dimethyl-3-oxo-7-propyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl]propanoate
SMILES CCCC1CC2=CC(=O)CCC2(C3C1C4CCC(C4(CC3)C)(CCC(=O)[O-])O)C.[K+]

Specifications

Quantity 50 mg
Formula Weight 440.66
Percent Purity >98%
Chemical Name or Material RU 28318, potassium salt
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.