Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ RU.521
GREENER_CHOICESDP

Catalog No. 790450 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
79-045-0 50 mg
79-041-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 79-045-0 Supplier Tocris Bioscience™ Supplier No. 7904/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective inhibitor of cyclic GMP-AMP synthase (cGAS)

RU.521 is a potent and selective cyclic GMP-AMP synthase (cGAS) inhibitor (Kd = 36.2 nM; IC50 values are 0.11 μM and 2.94 μM against m-cGAS and h-cGAS, respectively). RU.521 suppresses dsDNA-activated reporter activity but not 5'ppp-HP20 RNA or murine IFN-β-induced IFN-β1-dependent gene expression. RU.521 reduces constitutive expression of type I interferon in primary macrophages from a mouse model of Aicardi-Gouti?res syndrome. Pretreatment of chimeric antigen receptor (CAR) redirected T cells with RU.521 increases efficiency of virus-free gene editing by improving cell viability and CAR integration in cell line.

Specifications

Chemical Name or Material 3-[1-(6,7-Dichloro-1 H-benzimidazol-2-yl)-5-hydroxy-3-methyl-1 H-pyrazol-4-yl]-1(3 H)-isobenzofuranone
CAS 2262452-06-0
Quantity 50 mg
Target Synthase/Synthetase Inhibitors
Molecular Formula C19H12Cl2N4O3
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.