Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Supelco™ Rubber Stoppers
Shop All MilliporeSigma Supelco Products
Click to view available options
Product Type:
Rubber Stopper
Material:
Butyl Rubber
Rubber
Specifications
Specifications
| Color | Red |
| Material | Rubber |
| Product Type | Rubber Stopper |
| For Use With (Application) | Sealing Samples in Vials |
| Diameter (Metric) | 20 mm |
| Quantity | 100/Pk. |
| Hardness | 40-50 (Durometer (° Shore A)) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction