Learn More
Rubbermaid™ Polyethylene BRUTE™ Buckets
With wide pour spout
$65.00 - $508.00
Description
With wide pour spout
- Graduated in pints, quarts, and liters
- Resist acids, alkalis, paints, chemicals, and cleaning compounds
- Zinc-plated wire bail with grip