Learn More
Description
- Large-capacity buckets suit commercial applications
- 19-076-531 features foot pedal water evacuation; no lifting required
- Wringers are tested to exceed 50,000 wringing cycles, 58X the life of a traditional wringer
- Construction is premium tubular steel and structural web molded plastic
Order Info
Choose from systems with side-press or down-press wringers, 35 or 44 quart capacity. Optional Dirty Water Bucket is sold separately.
Specifications
Specifications
| Material | Polypropylene |
| Capacity (English) | 35 qt. |
| Color | Yellow |
| For Use With (Application) | For the Largest Commercial Cleaning jobs |
| Height (English) | 36.5 in. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.