Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rubella Virus E1 Monoclonal Antibody (S16D)
GREENER_CHOICE

Catalog No. PIMA518254
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA518254 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA518254 Supplier Invitrogen™ Supplier No. MA518254
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Purity is >90%.

Rubella virus was first isolated in 1962 by Parkman and Weller. Rubella virus is classified as a togavirus, genus Rubivirus. It is most closely related to group A arboviruses, such as Eastern and Western Equine Encephalitis viruses. It is an enveloped RNA virus, with a single antigenic type that does not cross-react with other members of the togavirus group. Rubella virus is relatively unstable and is inactivated by lipid solvents, trypsin, formalin, ultraviolet light, low pH, heat, and amantadine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rubella Virus E1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone S16D
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Host Species Mouse
Immunogen Rubella virus native protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.