Learn More
Invitrogen™ Rubella Virus Virions Polyclonal Antibody

Description
PA1-7242 detects the rubella virus. PA1-7242 has been successfully used in immunofluorescence procedures. The PA1-7242 immunogen is strain HPV77.
Specifications
Specifications
| Antigen | Rubella Virus Virions |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Host Species | Goat |
| Immunogen | Strain HPV77. |
| Purification Method | IgG Fraction |
| Quantity | 1 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.