Learn More
Description
RuBi-Glutamate is a ruthenium-bipyridine-trimethylphosphine caged glutamate that can be excited by visible wavelengths and releases glutamate after one- or two-photon excitation. Has high quantum efficiency and can be used at low concentrations, partly avoiding blockade of GABAergic transmission seen with other caged glutamate compounds. Displays high spatial resolution and generates excitatory responses in individual dendritic spines with physiological kinetics.
MNI-caged-L-glutamate (Cat. No. 1490) also available.
Specifications
Specifications
| CAS | 2417096-44-5 |
| Quantity | 10 mg |
| Product Line | Tocris |
| Product Type | RuBi-Glutamate |
| Molecular Weight (g/mol) | M.W. (Observed): 970.54 |
| Inhibitors | RuBi-Glutamate |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

