Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rubicon Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP130461JF549
Description
Rubicon Polyclonal antibody specifically detects Rubicon in Human samples. It is validated for Western BlotSpecifications
Rubicon | |
Polyclonal | |
Western Blot | |
Baron, Beclin 1-interacting protein, Beclin-1 Associated RUN Domain Containing Protein, KIAA0226, Rubicon, RUN And Cysteine Rich Domain Containing Beclin 1 Interacting Protein, RUN Domain And Cysteine-Rich Domain Containing, Beclin 1-Interacting Protein, Run Domain Beclin-1-Interacting And Cysteine-Rich Domain-Containing Protein, rundataxin, SCAR15 | |
The immunogen for this product maps to a region between residue 922 and 972 of human Rubicon using the numbering given in entry NP_055502.1 (GeneID 9711). | |
0.1 mL | |
Autophagy, Cancer, Cardiovascular Biology, Cell Biology, Endocrinology, Signal Transduction | |
9711 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction