Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rubidium, 99.75% (metals basis)

Catalog No. aa4421406
Click to view available options
Quantity:
5 g

CAS: 7440-17-7 | Rb | 85.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-17-7
Molecular Formula Rb
Molecular Weight (g/mol) 85.47
MDL Number MFCD00134055
InChI Key IGLNJRXAVVLDKE-UHFFFAOYSA-N
Synonym unii-mlt4718tjw, mlt4718tjw, rubidio, rubidium-103, rubidium-106, atom, rubidium, isotope of mass 103, rubidium, isotope of mass 106, un1423 dangerous when wet
PubChem CID 5357696
ChEBI CHEBI:33322
IUPAC Name rubidium
SMILES [Rb]

Specifications

Color Silver
Appearance Silver-colored solid
Density 1.532g/cm3
Quantity 5 g
Odor Odorless
Assay Percent Range 99.75% (Metals basis)
Chemical Name or Material Rubidium
What type of container is Rubidium 99.75% (metal basis) sold in?

Cat. No. 044214.xx Rubidium 99.75% (metal basis) is ampouled under Argon.

Cat. No. 010315.xx Rubidium 99.75% (metal basis) is vacuum sealed in a breakseal ampoule.

Hazard Category H260-H314-H335
Precautionary Statement P223-P231+P232-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c
DOTInformation DOT Class: 4.3, Packaging Group: I
EINECSNumber 231-126-6
RTECSNumber VL8500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.