Learn More
Rubidium acetate, 99.8% (metals basis)
Description
Rubidium acetate has been reported to modify the acid/base character of the catalyst supports like silica, alumina, and titania. It can be used in the preparation of nanosized rubidium ferrite, and as a promoter on the palladium-catalyzed synthesis of vinyl acetate in heterogeneous gas reaction. It is used in medical and in research laboratories.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 246°C |
Quantity | 50 g |
Sensitivity | Hygroscopic |
Solubility Information | Slightly soluble in water. |
Formula Weight | 144.52 |
Percent Purity | 99.8% |
Physical Form | Crystalline |
Assay | (metals basis) |
Chemical Name or Material | Rubidium acetate |
Safety and Handling
EINECSNumber | 209-255-4 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.