Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rubidium chloride, 99.8+%, (trace metal basis), ACROS Organics™

Click to view available options
Quantity:
10g
50g
250g
Packaging:
Glass bottle
7791-11-9
ClRb
120.92
FGDZQCVHDSGLHJ-UHFFFAOYSA-M
rubidium(1+) chloride
[Cl-].[Rb+]
7791-11-9
ClRb
120.92
FGDZQCVHDSGLHJ-UHFFFAOYSA-M
rubidium(1+) chloride
[Cl-].[Rb+]
For ultra trace analysis
Assay Percent Range 99.8+%
Linear Formula RbCl
Merck Index 15,8417
Solubility Information Solubility in water: 910g/L (20 c). Other solubilities: 1g/0.7mL boiling water,practically insoluble in alcohols
Formula Weight 120.92
Physical Form Crystals
Percent Purity 99.8+%
Grade Trace Metal Basis
Packaging Glass bottle
Solubility Solubility in water: 910g/L (20 c). Other solubilities: 1g/0.7mL boiling water, practically insoluble in alcohols
Total Trace Metal Impurities 2000.0ppm max
Boiling Point 1390.0°C
Color White
Melting Point 718.0°C
Quantity 250g
Chemical Name or Material Rubidium chloride
Show More Show Less

EINECSNumber : 232-240-9