Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rubidium chloride, 99+%

Catalog No. p-3735328
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle

CAS: 7791-11-9 | ClRb | 120.92 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7791-11-9
Molecular Formula ClRb
Molecular Weight (g/mol) 120.92
MDL Number MFCD00011187
InChI Key FGDZQCVHDSGLHJ-UHFFFAOYSA-M
Synonym rubidium chloride, rubidium chloride rbcl, rubidium monochloride, rbcl, rubidium 1+ chloride, rubidium 1+ ion chloride, unii-n3shc5273s, rubidiumchloride, rubidium-chlorid, rubidium-chloride
PubChem CID 62683
ChEBI CHEBI:78672
SMILES [Cl-].[Rb+]

Specifications

Boiling Point 1390.0°C
Melting Point 718.0°C
Color White
Physical Form Crystalline Powder and/or Crystals
Quantity 25 g
Assay Percent Range 99+%
Linear Formula RbCl
Merck Index 15, 8417
Solubility Information Solubility in water: 910g/L (20 c). Other solubilities: 1g/0.7mL boiling water, practically insoluble in alcohols
IUPAC Name rubidium(1+);chloride
Formula Weight 120.92
Percent Purity ≥99%
Packaging Glass bottle
Density 2.8g/mL
Chemical Name or Material Rubidium chloride
Show More Show Less
EINECSNumber 232-240-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.