Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rubidium hydroxide, 50% w/w aq. soln., 99.6+% (metals basis)

Catalog No. p-7048447
Click to view available options
Quantity:
10 g
50 g

CAS: 1310-82-3 | HORb | 102.48 g/mol

Rubidium Hydroxide is used to give fireworks a violet color. It is a notable ntermediate in the synthesis of other rubidium compounds. It is used in photography and storage batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-82-3
Molecular Formula HORb
Molecular Weight (g/mol) 102.48
MDL Number MFCD00011194
InChI Key CPRMKOQKXYSDML-UHFFFAOYSA-M
Synonym rubidium hydroxide, rubidium hydroxide rb oh, rboh, rubidium 1+ hydroxide, rubidium hydroxide solution, rubidiumol, rubidium hydoxide, rubidium hydroxide sup 81 rb, rubidium monohydroxide, rb.ho
PubChem CID 62393
ChEBI CHEBI:32108
IUPAC Name rubidium(1+);hydroxide
SMILES [OH-].[Rb+]

Specifications

Color Yellow
Physical Form Liquid
Quantity 10 g
Linear Formula RbOH
UN Number UN2677
Merck Index 14,8285
Solubility Information Soluble in ethanol and water.
Formula Weight 102.48 (Anhydrous)
Percent Purity ≥99.6%
Concentration or Composition (by Analyte or Components) 50% w/w aq. soln.
Assay (metals basis)
Chemical Name or Material Rubidium hydroxide
Show More Show Less
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H314-H302
Causes severe skin burns and eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: RUBIDIUM HYDROXIDE SOLUTION
EINECSNumber 215-186-0
RTECSNumber VL8750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.