Alfa Aesar™ Rubidium nitrate, 99.8% (metals basis)

Manufacturer: Alfa Aesar™ 1288909

Quantity 10g

 View more versions of this product

Catalog No. AA1288909

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13126-12-0
    Formula Weight 147.47
    MDL Number MFCD00011193
    UN Number UN1477
    Chemical Name or Material Rubidium nitrate

    Specifications

    Formula Weight 147.47
    Health Hazard 3 GHS P Statement
    P221-P210-P220-P280-P305+P351+P338-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Keep/Store away from clothing/combustible materials.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    UN Number UN1477
    Health Hazard 2 GHS H Statement
    H272-H315-H319
    May intensify fire
    oxidizer.
    Causes skin irritation.
    Causes serious eye irritation.
    Density 3.11g/mL
    TSCA TSCA
    Physical Form Crystalline
    Melting Point 310°C
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.
    Refractive Index 1.52
    Sensitivity Hygroscopic
    MDL Number MFCD00011193
    CAS 13126-12-0
    Quantity 10g
    RTECS Number QV0900000
    Color White
    Chemical Name or Material Rubidium nitrate
    Assay 99.8%, (metals basis)

    GHS H Statement
    H272-H315-H319
    May intensify fire
    oxidizer.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P221-P210-P220-P280-P305+P351+P338-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Keep/Store away from clothing/combustible materials.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit