Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rubidium, plasma standard solution, Specpure™ Rb 1000μg/mL

Catalog No. p-4824910
Click to view available options
Quantity:
50 mL
100 mL

CAS: 584-09-8 | CO3Rb2 | 230.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 584-09-8, 7697-37-2, 7732-18-5
Molecular Formula CO3Rb2
Molecular Weight (g/mol) 230.94
MDL Number MFCD00011188
InChI Key WPFGFHJALYCVMO-UHFFFAOYSA-L
PubChem CID 11431
IUPAC Name dirubidium(1+) carbonate
SMILES [Rb+].[Rb+].[O-]C([O-])=O

Specifications

Vapor Pressure 23 hPa (17mm Hg) at 20°C
Physical Form Liquid
Color Colorless
Quantity 50 mL
Concentration or Composition (by Analyte or Components) Nitric acid: 5.0%; Rubidium carbonate: 0.14%; Water: 94.86%
Chemical Name or Material Rubidium
Hazard Category H290-H314-H335
Precautionary Statement P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
DOTInformation Hazard Class: 8, Packing Group: III
EINECSNumber 231-714-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.