Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rubrene, 99%, ACROS Organics™

Click to view available options
Quantity:
500mg
1g
Packaging:
Glass bottle
517-51-1
C42H28
532.67
MFCD00003703
YYMBJDOZVAITBP-UHFFFAOYSA-N
rubrene,5,6,11,12-tetraphenylnaphthacene,rubren,naphthacene, 5,6,11,12-tetraphenyl,rubrene, powder,acmc-209kvj,5,11,12-tetraphenyltetracene,5,11,12-tetraphenylnaphthacene,naphthacene,6,11,12-tetraphenyl
68203
5,6,11,12-tetraphenyltetracene
C1=CC=C(C=C1)C2=C3C=CC=CC3=C(C4=C(C5=CC=CC=C5C(=C24)C6=CC=CC=C6)C7=CC=CC=C7)C8=CC=CC=C8
517-51-1
C42H28
532.67
MFCD00003703
YYMBJDOZVAITBP-UHFFFAOYSA-N
rubrene,5,6,11,12-tetraphenylnaphthacene,rubren,naphthacene, 5,6,11,12-tetraphenyl,rubrene, powder,acmc-209kvj,5,11,12-tetraphenyltetracene,5,11,12-tetraphenylnaphthacene,naphthacene,6,11,12-tetraphenyl
68203
5,6,11,12-tetraphenyltetracene
C1=CC=C(C=C1)C2=C3C=CC=CC3=C(C4=C(C5=CC=CC=C5C(=C24)C6=CC=CC=C6)C7=CC=CC=C7)C8=CC=CC=C8
Melting Point 330°C to 335°C
Quantity 500mg
Packaging Glass bottle
Infrared Spectrum Authentic
Color Red
Assay Percent Range 98.5% min. (HPLC)
Beilstein 05, II, 726
Solubility Information Solubility in water: practically insoluble
Formula Weight 532.67
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material Rubrene, 99%
Show More Show Less

EINECSNumber : 208-242-0

TSCA : TSCA